accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 618-45-1

H3CCH3HO

CAS RN 618-45-1

IUPAC Name: 3-(propan-2-yl)phenol
Synonyms: M-isopropylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: VLJSLTNSFSOYQR-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results