CAS RN 61733-01-5
CAS RN 61733-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Melphalan (>90%), TRC
CAS: 61733-01-5 Molecular Formula: C13 H19 Cl N2 O3 Molecular Weight (g/mol): 286.75 Synonym: L-Phenylalanine, 4-[(2-chloroethyl)(2-hydroxyethyl)amino]-,4-[(2-Chloroethyl)(2-hydroxyethyl)amino]-L-phenylalanine,Hydroxymelphalan,(S)-2-Amino-3-(4-((2-chloroethyl)(2-hydroxyethyl)amino)phenyl)propanoic acid,Monohydroxy Melphalan,Monohydroxymelphalan IUPAC Name: (2S)-2-amino-3-[4-[2-chloroethyl(2-hydroxyethyl)amino]phenyl]propanoic acid SMILES: N[C@@H](Cc1ccc(cc1)N(CCO)CCCl)C(=O)O