accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6160-65-2

SNNNN

CAS RN 6160-65-2

IUPAC Name: 1-(1H-imidazole-1-carbothioyl)-1H-imidazole
Synonyms: 1-(imidazole-1-carbothioyl)imidazole
Molecular Weight (g/mol): 178.21
Molecular Formula: C7H6N4S
InChi Key: RAFNCPHFRHZCPS-UHFFFAOYSA-N
SMILES: S=C(N1C=CN=C1)N1C=CN=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)