Learn More
1,1’-Thiocarbonyldiimidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1'-Thiocarbonyldiimidazole |
| CAS | 6160-65-2 |
| Molecular Formula | C7 H6 N4 S |
| Quantity | 5 g |
| Synonym | di-1H-Imidazol-1-ylmethanethione, 1,1'-Carbonothioylbis-1H-imidazole, ,'1,1'-(Thiocarbonyl)diimidazole, 1,1'-(Thiocarbonyl)bis[imidazole], 1,1'-(Thiocarbonyl)diimidazole, 1-(1H-Imidazol-1-ylcarbothioyl)-1H-imidazole, Diimidazol-1-ylmethanthione, N,N'-Thiocarbonyldiimidazole, NSC 141692, Thiocarbonylbisimidazole, Thiocarbonyldiimidazole, TCDI |
| InChI Formula | InChI=1S/C7H6N4S/c12-7(10-3-1-8-5-10)11-4-2-9-6-11/h1-6H |
| SMILES | S=C(n1ccnc1)n2ccnc2 |
| IUPAC Name | di(imidazol-1-yl)methanethione |
| Molecular Weight (g/mol) | 178.214 |
| Formula Weight | 178.031 |