CAS RN 609-89-2
CAS RN 609-89-2
IUPAC Name:
2,4-dichloro-6-nitrophenol
Synonyms:
2,4-dichloro-6-nitrophenol
Molecular Weight (g/mol):
207.99
Molecular Formula:
C6H3Cl2NO3
InChi Key:
LYPMXMBQPXPNIQ-UHFFFAOYSA-N
SMILES:
OC1=C(Cl)C=C(Cl)C=C1[N+]([O-])=O
1
–
1
of
1
results
1
2,4-Dichloro-6-nitrophenol, 98+%
CAS: 609-89-2 Molecular Formula: C6H3Cl2NO3 Molecular Weight (g/mol): 207.994 MDL Number: MFCD00007101 InChI Key: LYPMXMBQPXPNIQ-UHFFFAOYSA-N Synonym: 2,4-dichloro-6-nitrophenol,phenol, 2,4-dichloro-6-nitro,2,4-dichlor-6-nitrofenol,4,6-dichloro-2-nitrophenol,2,4-dichlor-6-nitrofenol czech,pubchem22918,acmc-1aunw,wln: wnr bq cg eg,2-nitro-4,6-dichlorophenol PubChem CID: 11871 SMILES: C1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl