accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONCH3O

CAS RN 59857-86-2

IUPAC Name: methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
Synonyms: methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: JFZRRXGXEHUJCI-UHFFFAOYNA-N
SMILES: COC(=O)C1CN(C)C(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results