accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 598-30-1

IUPAC Name: lithium(1+) butan-2-ide
Synonyms: sec-butyllithium
Molecular Weight (g/mol): 64.06
Molecular Formula: C4H9Li
InChi Key: WGOPGODQLGJZGL-UHFFFAOYSA-N
SMILES: [Li+].CC[CH-]C
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results