accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 59382-59-1

CH3OOH3CNOO

CAS RN 59382-59-1

IUPAC Name: methyl 2-methyl-3-nitrobenzoate
Synonyms: methyl 2-methyl-3-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: CRZGFIMLHZTLGT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1C)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results