accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3P

CAS RN 5931-53-3

IUPAC Name: (2-methylphenyl)diphenylphosphane
Synonyms: (2-methylphenyl)diphenylphosphane
Molecular Weight (g/mol): 276.32
Molecular Formula: C19H17P
InChi Key: MLBZLJCMHFCTQM-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results