Learn More
Diphenyl(o-tolyl)phosphine
Product Code. 15483697
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483697 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15483697 Supplier Thermo Scientific Alfa Aesar Supplier No. H27503.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 5931-53-3 | C19H17P | 276.319 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 5931-53-3
Melting Point 67°C to 70°C
Molecular Formula C19H17P
MDL Number MFCD00008515
Quantity 1 g
Synonym diphenyl o-tolyl phosphine, 2-methylphenyl diphenylphosphine, diphenyl-o-tolylphosphine, o-tolyldiphenylphosphine, 2-methylphenyl diphenylphosphane, 2-methylphenyl diphenyl phosphine, diphenyltolylphosphine, acmc-1bwkt, diphenyl 2-methylphenyl phosphine
InChI Key MLBZLJCMHFCTQM-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3
IUPAC Name (2-methylphenyl)-diphenylphosphane
Molecular Weight (g/mol) 276.319
PubChem CID 80040
Formula Weight 276.32
Chemical Name or Material Diphenyl(o-tolyl)phosphine