CAS RN 593-60-2
CAS RN 593-60-2
IUPAC Name:
bromoethene
Synonyms:
vinyl bromide bromure de vinyle vinylbromid C2H3br
Molecular Weight (g/mol):
106.95
Molecular Formula:
C2H3Br
InChi Key:
INLLPKCGLOXCIV-UHFFFAOYSA-N
SMILES:
BrC=C
1
–
2
of
2
results
1
Vinyl Bromide 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Vinyl bromide, 1M solution in THF, AcroSeal™
CAS: 593-60-2 | C2H3Br | 106.95 g/mol