accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNCH3ON

CAS RN 59-26-7

IUPAC Name: N,N-diethylpyridine-3-carboxamide
Synonyms: corvin nikethamide (product) nicethamidum nicotinic diethylamide nisetamide dietilamide-carbopiridina β-pyrimidum cardamine diaethylamidum acidi nicotinici diethylamide, nicotinic
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: NCYVXEGFNDZQCU-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)