accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COSOONH2

CAS RN 58734-57-9

IUPAC Name: 3-methoxybenzene-1-sulfonamide
Synonyms: benzenesulfonamide, 3-methoxy-
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: VBKIEQKVSHDVGH-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results