CAS RN 579492-81-2
CAS RN 579492-81-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
MedChemExpress Bax inhibitor peptide V5
MedChemExpress Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, used for cancer treatment.
2
MedChemExpress Bax inhibitor peptide V5
MedChemExpress Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, used for cancer treatment.
3
MedChemExpress Bax inhibitor peptide V5
MedChemExpress Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, used for cancer treatment.
4
MedChemExpress Bax inhibitor peptide V5
MedChemExpress Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, used for cancer treatment.