Learn More
MedChemExpress Bax inhibitor peptide V5
Product Code. 16421808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16421808
Brand: MedChemExpress HYP0081
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, used for cancer treatment.
Specifications
Purity 98.12%
CAS 579492-81-2
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 586.79 g/mol
Product Type Reference compound
Quantity 5 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (170.42 mM) ∣H2O : 100 mg/mL (170.42 mM∣Need ultrasonic)
Synonym BIP-V5∣ BAX Inhibiting Peptide V5