CAS RN 57637-97-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Aminomethyl phosphonic acid N-acetyl 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
N-Acetylaminomethylphosphoric Acid (>90%), TRC
CAS: 57637-97-5 Molecular Formula: C3 H8 N O4 P Molecular Weight (g/mol): 153.07 Synonym: Phosphonic acid, (acetamidomethyl)- (6CI),Phosphonic acid, [(acetylamino)methyl]- (9CI),P-[(Acetylamino)methyl]phosphonic acid,N-Acetylaminomethylphosphonic acid,Acetamidomethylphosphonic Acid IUPAC Name: acetamidomethylphosphonic acid SMILES: CC(=O)NCP(=O)(O)O