CAS RN 56817-09-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Chloro-imidazo[1,2-C]pyrimidin-5-ol, TRC
CAS: 56817-09-5 Molecular Formula: C6H4N3OCl Molecular Weight (g/mol): 169.56 Synonym: Imidazo[1,2-c]pyrimidin-5(6H)-one, 7-chloro- (9CI, ACI),7-Chloroimidazo[1,2-c]pyrimidin-5(6H)-one (ACI),7-Chloro-imidazo[1,2-c]pyrimidin-5-ol IUPAC Name: 7-chloro-6H-imidazo[1,2-c]pyrimidin-5-one SMILES: ClC1=Cc2nccn2C(=O)N1