accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5675-51-4

HOOH

CAS RN 5675-51-4

IUPAC Name: dodecane-1,12-diol
Synonyms: 1,12-dodecanediol
Molecular Weight (g/mol): 202.34
Molecular Formula: C12H26O2
InChi Key: GHLKSLMMWAKNBM-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)