Learn More
1,12-Dodecanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,12-Dodecanediol |
| CAS | 5675-51-4 |
| Molecular Formula | C12 H26 O2 |
| Quantity | 25 g |
| Synonym | 1,12-Dodecanediol (6CI, 7CI, 8CI, 9CI, ACI), 1,12-Dihydroxydodecane, D 0969, Dodecamethylene glycol, n-Dodecane-1,12-diol, NSC 81250, α,ω-Dodecanediol |
| InChI Formula | InChI=1S/C12H26O2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h13-14H,1-12H2 |
| SMILES | OCCCCCCCCCCCCO |
| IUPAC Name | dodecane-1,12-diol |
| Molecular Weight (g/mol) | 202.33 |
| Formula Weight | 202.1933 |