CAS RN 5674-02-2
IUPAC Name:
(2-methylpropyl)magnesiumylium chloride
Synonyms:
(2-methylpropyl)magnesiumylium chloride
Molecular Weight (g/mol):
116.87
Molecular Formula:
C4H9ClMg
InChi Key:
YUPZOKFCNKSJTR-UHFFFAOYSA-M
SMILES:
[Cl-].CC(C)C[Mg+]
1
–
2
of
2
results
2
Isobutylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
CAS: 5674-02-2 Molecular Weight (g/mol): 116.87 MDL Number: MFCD00000471 ChEBI: CHEBI:75272