accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ZnCH3H3C

CAS RN 557-20-0

IUPAC Name: diethylzinc
Synonyms: diethylzinc
Molecular Weight (g/mol): 123.5
Molecular Formula: C4H10Zn
InChi Key: IPSRAFUHLHIWAR-UHFFFAOYSA-N
SMILES: [Zn++].C[CH2-].C[CH2-]
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results