accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OONH(S)H2NHOO

CAS RN 55533-24-9

IUPAC Name: (2S)-3-(4-aminophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-3-(4-aminophenyl)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: NDMVQEZKACRLDP-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results