accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 551-92-8

CH3NCH3NNOO

CAS RN 551-92-8

IUPAC Name: 1,2-dimethyl-5-nitro-1H-imidazole
Synonyms: dimetridazole
Molecular Weight (g/mol): 141.13
Molecular Formula: C5H7N3O2
InChi Key: IBXPYPUJPLLOIN-UHFFFAOYSA-N
SMILES: CN1C(C)=NC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Certifications/Compliance 
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)