accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 547-57-9

NHNOOHSOOOHNa(E/Z)

CAS RN 547-57-9

IUPAC Name: 4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonic acid sodium
Synonyms: 4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzenesulfonic acid sodium
Molecular Weight (g/mol): 317.27
Molecular Formula: C12H10N2NaO5S
InChi Key: YOGUPPQYRWYKGT-UHFFFAOYSA-N
SMILES: [Na].OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)