accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 54143-56-5

CH3OHOFFFOOFFFOHNHN

CAS RN 54143-56-5

IUPAC Name: N-[(piperidin-2-yl)methyl]-2,5-bis(2,2,2-trifluoroethoxy)benzamide; acetic acid
Synonyms: acetic acid; flecainide
Molecular Weight (g/mol): 474.4
Molecular Formula: C19H24F6N2O5
InChi Key: RKXNZRPQSOPPRN-UHFFFAOYNA-N
SMILES: CC(O)=O.FC(F)(F)COC1=CC=C(OCC(F)(F)F)C(=C1)C(=O)NCC1CCCCN1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results