accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 540-08-9

H3COCH3

CAS RN 540-08-9

IUPAC Name: heptadecan-9-one
Synonyms: 9-heptadecanone
Molecular Weight (g/mol): 254.46
Molecular Formula: C17H34O
InChi Key: WTJKUFMLQFLJOT-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)CCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)