Learn More
9-Heptadecanone, TRC
Specifications
Specifications
| Chemical Name or Material | 9-Heptadecanone |
| CAS | 540-08-9 |
| Molecular Formula | C17H34O |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C17H34O/c1-3-5-7-9-11-13-15-17(18)16-14-12-10-8-6-4-2/h3-16H2,1-2H3 |
| SMILES | CCCCCCCCC(=O)CCCCCCCC |
| IUPAC Name | heptadecan-9-one |
| Molecular Weight (g/mol) | 254.45 |
| Formula Weight | 254.261 |