accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOClBrBr

CAS RN 5278-95-5

IUPAC Name: 2,2-dibromo-2-chloroacetic acid
Synonyms: dibromochloroacetic acid
Molecular Weight (g/mol): 252.29
Molecular Formula: C2HBr2ClO2
InChi Key: UCZDDMGNCJJAHK-UHFFFAOYSA-N
SMILES: OC(=O)C(Cl)(Br)Br
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results