accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHNNHNNS

CAS RN 52431-78-4

IUPAC Name: 1-(4-hydroxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms: 1-(4-hydroxyphenyl)-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol): 194.21
Molecular Formula: C7H6N4OS
InChi Key: MOXZSKYLLSPATM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N1NN=NC1=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results