accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 52374-18-2

H3CHNSOONOO

CAS RN 52374-18-2

IUPAC Name: N-hexyl-4-nitrobenzene-1-sulfonamide
Synonyms: N-hexyl-4-nitrobenzenesulfonamide
Molecular Weight (g/mol): 286.35
Molecular Formula: C12H18N2O4S
InChi Key: HMKTZTGMSGRPHW-UHFFFAOYSA-N
SMILES: CCCCCCNS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results