accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 52009-64-0

NH2NH2N

CAS RN 52009-64-0

IUPAC Name: 6-phenylphenanthridine-3,8-diamine
Synonyms: 6-phenylphenanthridine-3,8-diamine
Molecular Weight (g/mol): 285.35
Molecular Formula: C19H15N3
InChi Key: CPNAVTYCORRLMH-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=C1)N=C(C1=CC=CC=C1)C1=CC(N)=CC=C21
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)