Learn More
6-Phenylphenathridine-3,8-diamine, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Phenylphenathridine-3,8-diamine |
| CAS | 52009-64-0 |
| Molecular Formula | C19 H15 N3 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C19H15N3/c20-13-6-8-15-16-9-7-14(21)11-18(16)22-19(17(15)10-13)12-4-2-1-3-5-12/h1-11H,20-21H2 |
| SMILES | Nc1ccc2c(c1)nc(c3ccccc3)c4cc(N)ccc24 |
| IUPAC Name | 6-phenylphenanthridine-3,8-diamine |
| Molecular Weight (g/mol) | 285.34 |
| Formula Weight | 285.1266 |