accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51896-27-6

CH3ClSNH2OO

CAS RN 51896-27-6

IUPAC Name: 3-chloro-4-methylbenzene-1-sulfonamide
Synonyms: 3-chloro-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 205.66
Molecular Formula: C7H8ClNO2S
InChi Key: YAUUWNCFVUUNCM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results