accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51871-62-6

H3CH3CCH3OONH(S)HOO

CAS RN 51871-62-6

IUPAC Name: (3S)-3-{[(tert-butoxy)carbonyl]amino}-4-phenylbutanoic acid
Synonyms: (3S)-3-[(tert-butoxycarbonyl)amino]-4-phenylbutanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: ACKWQHCPHJQANL-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC(O)=O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)