accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOHHO

CAS RN 51863-60-6

IUPAC Name: 1-(3,5-dihydroxyphenyl)ethan-1-one
Synonyms: 1-(3,5-dihydroxyphenyl)ethanone
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: WQXWIKCZNIGMAP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)