Learn More
3',5'-Dihydroxyacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 3',5'-Dihydroxyacetophenone |
| CAS | 51863-60-6 |
| Molecular Formula | C8H8O3 |
| Quantity | 250 g |
| Synonym | 1-(3,5-Dihydroxyphenyl)ethanone, 3,5-Dihydroxyacetophenone, 3',5'-Dihydroxyphenyl Methyl Ketone |
| InChI Formula | InChI=1S/C8H8O3/c1-5(9)6-2-7(10)4-8(11)3-6/h2-4,10-11H,1H3 |
| SMILES | CC(=O)c1cc(O)cc(O)c1 |
| Molecular Weight (g/mol) | 152.15 |
| Formula Weight | 152.0473 |