CAS RN 50907-21-6
IUPAC Name:
5-(4-fluorophenyl)-2H-1,2,3,4-tetrazole
Synonyms:
5-(4-fluorophenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol):
164.14
Molecular Formula:
C7H5FN4
InChi Key:
DTAOERLWDORJFR-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C1=NNN=N1
1
–
1
of
1
results
1
5-(4-Fluorophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals
CAS: 50907-21-6 Molecular Formula: C7H5FN4 Molecular Weight (g/mol): 164.143 MDL Number: MFCD00478201 InChI Key: DTAOERLWDORJFR-UHFFFAOYSA-N Synonym: 5-4-fluorophenyl-1h-tetrazole,5-4-fluorophenyl-2h-tetrazole,5-4-fluoro-phenyl-2h-tetrazole,5-4-fluorophenyl-1h-1,2,3,4-tetrazole,5-4-fluorophenyl-2h-1,2,3,4-tetrazole,5-4-fluorophenyl-1h-1,2,3,4-tetraazole,acmc-20am8i,4-fluorophenyl tetrazole,cambridge id 5612113,5-4-fluorophenyl tetrazole PubChem CID: 766140 IUPAC Name: 5-(4-fluorophenyl)-2H-tetrazole SMILES: C1=CC(=CC=C1C2=NNN=N2)F