accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50892-23-4

CH3HNClNSOHONH3C

CAS RN 50892-23-4

IUPAC Name: 2-({4-chloro-6-[(2,3-dimethylphenyl)amino]pyrimidin-2-yl}sulfanyl)acetic acid
Synonyms: pirinixic acid
Molecular Weight (g/mol): 323.8
Molecular Formula: C14H14ClN3O2S
InChi Key: SZRPDCCEHVWOJX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(NC2=CC(Cl)=NC(SCC(O)=O)=N2)=C1C
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results