Learn More
Pirinixic acid, MedChemExpress
Product Code. 16477248
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
50mg
Product Code. 16477248
Brand: Medchem Express HY16995
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Pirinixic acid (Wy-14643) is a potent agonist of PPARα, with EC50s of 0.63 μM, 32 μM for murine PPARα and PPARγ, and 5.0 μM, 60 μM, 35 μM for human PPARα, PPARγ and PPARδ, respectively.
Pirinixic acid (Wy-14643) is a potent agonist of PPARα, with EC50s of 0.63 μM, 32 μM for murine PPARα and PPARγ, and 5.0 μM, 60 μM, 35 μM for human PPARα, PPARγ and PPARδ, respectively.
Specifications
Chemical Name or Material Pirinixic acid
Physical Form Powder
CAS 50892-23-4
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H14ClN3O2S
For Use With (Application) COVID-19-anti-virus
Synonym Wy-14643
Solubility Information DMSO : 100 mg/mL (308.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)CSC1=NC(NC2=CC=CC(C)=C2C)=CC(Cl)=N1
Molecular Weight (g/mol) 323.8
Formula Weight 323.8
Percent Purity 98.72%
Grade Research