accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50703-06-5

HClCH3OOSHN

CAS RN 50703-06-5

IUPAC Name: hydrogen methyl 1,3-thiazolidine-4-carboxylate chloride
Synonyms: hydrogen methyl 1,3-thiazolidine-4-carboxylate chloride
Molecular Weight (g/mol): 183.65
Molecular Formula: C5H10ClNO2S
InChi Key: YEFOSJYXVLNYSL-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C1CSCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results