accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50528-80-8

IUPAC Name: 1,3,5-trifluoro-2-isocyanatobenzene
Synonyms: 1,3,5-trifluoro-2-isocyanatobenzene
Molecular Weight (g/mol): 173.09
Molecular Formula: C7H2F3NO
InChi Key: XORSGSHLSDFOHP-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(N=C=O)C(F)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results