CAS RN 50-90-8
CAS RN 50-90-8
IUPAC Name:
5-chloro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
5-chloro-2'-deoxyuridine
Molecular Weight (g/mol):
262.65
Molecular Formula:
C9H11ClN2O5
InChi Key:
NJCXGFKPQSFZIB-RRKCRQDMSA-N
SMILES:
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Cl)C(=O)NC1=O
1
–
3
of
3
results
2
5-Chloro-2'-deoxyuridine, MP Biomedicals™
CAS: 50-90-8 Molecular Formula: C9H11ClN2O5 Molecular Weight (g/mol): 262.646 InChI Key: NJCXGFKPQSFZIB-XVMARJQXSA-N Synonym: 5-chloro-2'-deoxyuridine PubChem CID: 13831089 IUPAC Name: 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione SMILES: C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O
3
5-Chloro-2'-deoxyuridine, MedChemExpress
MedChemExpress 5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.