CAS RN 50-86-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Acetyl-4-aminosalicylic Acid, TRC
CAS: 50-86-2 Molecular Formula: C9 H9 N O4 Molecular Weight (g/mol): 195.17 Synonym: 4-(Acetylamino)-2-hydroxybenzoic Acid,Metoclopramide Imp. H (EP) IUPAC Name: 4-acetamido-2-hydroxybenzoic acid SMILES: CC(=O)Nc1ccc(C(=O)O)c(O)c1
2
4-(Acetylamino)-2-hydroxybenzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.