accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3H3CH3C

CAS RN 488-23-3

IUPAC Name: 1,2,3,4-tetramethylbenzene
Synonyms: 1,2,3,4-tetramethylbenzene prehnitene prehnitol
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: UOHMMEJUHBCKEE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(C)=C1C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results