CAS RN 487-79-6
CAS RN 487-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Kainic acid (synthetic)
Kainate agonist; excitant and neurotoxin
2
Kainic acid, MedChemExpress
MedChemExpress Kainic acid is a potent agonist at excitatory amino acid receptor subtypes in the CNS.