Learn More
Kainic acid, MedChemExpress
Product Code. 16469278
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16469278
Brand: Medchem Express HYN2309
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Kainic acid is a potent agonist at excitatory amino acid receptor subtypes in the CNS.
Kainic acid is a potent agonist at excitatory amino acid receptor subtypes in the CNS.
Specifications
Chemical Name or Material Kainic acid
CAS 487-79-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H15NO4
Quantity 10 mg
Solubility Information H2O : 25 mg/mL (117.24 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)C[C@@H]1[C@@H](C(O)=O)NC[C@@H]1C(C)=C
Molecular Weight (g/mol) 213.23
Formula Weight 213.23
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation