accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 48068-25-3

(R)OHO(S)HNH3COOOCH3CH3CH3H3C

CAS RN 48068-25-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-methoxybutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-methoxybutanoic acid
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: VWSUOKFUIPMDDX-UHFFFAOYNA-N
SMILES: COC(C)C(NC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)