accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOOF

CAS RN 458-09-3

IUPAC Name: 2-(3-fluoro-4-hydroxyphenyl)acetate
Synonyms: 2-(3-fluoro-4-hydroxyphenyl)acetate
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H6FO3
InChi Key: YRFBZAHYMOSSGX-UHFFFAOYSA-M
SMILES: OC1=CC=C(CC([O-])=O)C=C1F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)