accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COCH3

CAS RN 4485-09-0

IUPAC Name: nonan-4-one
Synonyms: 4-nonanone
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: TYBCSQFBSWACAA-UHFFFAOYSA-N
SMILES: CCCCCC(=O)CCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)