accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 443-69-6

FHNOO

CAS RN 443-69-6

IUPAC Name: 5-fluoro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-fluoro-1H-indole-2,3-dione
Molecular Weight (g/mol): 165.12
Molecular Formula: C8H4FNO2
InChi Key: GKODDAXOSGGARJ-UHFFFAOYSA-N
SMILES: FC1=CC=C2NC(=O)C(=O)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results